Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197104.1 | NP_001184033.1 | 13 - 14 | 4719 | 108 | |
NM_005933.3 | NP_005924.2 | 13 - 14 | 4719 | 108 | ||
XM_006718839.3 | XP_006718902.2 | 13 - 14 | 2194 | 108 | ||
XM_011542829.2 | XP_011541131.1 | 14 - 15 | 4810 | 108 | ||
XM_011542830.2 | XP_011541132.1 | 14 - 15 | 4807 | 108 | ||
XM_011542831.2 | XP_011541133.1 | 14 - 15 | 4810 | 108 | ||
XM_011542833.2 | XP_011541135.1 | 14 - 15 | 2293 | 108 | ||
GenBank mRNA | AB209508.1 | - | 11 - 12 | 2263 | 108 | |
AF232001.1 | - | 7 - 8 | 790 | 108 | ||
D14540.1 | - | 13 - 14 | 4719 | 108 | ||
L01986.1 | - | 7 - 8 | 750 | 108 | ||
L04284.1 | - | 13 - 14 | 4696 | 108 | ||
L04731.1 | - | 13 - 14 | 4510 | 108 | ||
S78570.1 | - | 6 - 7 | 832 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |