Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003244.hg.1 |
TC04000221.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003244.hg.1 |
ENST00000261438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003244.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003244.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012824.hg.1 |
ENST00000521506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003238.hg.1 |
TC04000221.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003238.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003243.hg.1 |
TC04000221.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003243.hg.1 |
ENST00000261438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003243.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003243.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003242.hg.1 |
TC04000221.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003242.hg.1 |
ENST00000261438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003242.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003242.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000482150.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000482150.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000482150.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000482150.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149637.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149637.hg.1 |
ENST00000482150.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149637.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149637.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149637.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149637.hg.1 |
ENST00000482150.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149637.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149637.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149637.hg.1 |
TC0400007264.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000482150.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000482150.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000482150.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000482150.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007264.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012824.hg.1 |
ENST00000521506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149636.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149636.hg.1 |
ENST00000482150.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149636.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149636.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149636.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149636.hg.1 |
ENST00000482150.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149636.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149636.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149636.hg.1 |
TC0400007264.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400149631.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149631.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149631.hg.1 |
TC0400007264.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400149635.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149635.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149635.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149635.hg.1 |
ENST00000261438.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149635.hg.1 |
ENST00000514033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149635.hg.1 |
NM_016531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149635.hg.1 |
TC0400007264.hg.1 |
Clariom D TC |