Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017934.5 | NP_060404.3 | 22 - 23 | 2765 | 72 | |
XM_005248729.4 | XP_005248786.1 | 22 - 23 | 2762 | 72 | ||
XM_011535918.2 | XP_011534220.1 | 19 - 20 | 2421 | 72 | ||
XM_011535919.1 | XP_011534221.1 | 22 - 23 | 2765 | 72 | ||
XM_017010989.1 | XP_016866478.1 | 24 - 25 | 2907 | 72 | ||
XM_017010990.1 | XP_016866479.1 | 21 - 22 | 2505 | 72 | ||
GenBank mRNA | AJ303102.1 | - | 22 - 23 | 2552 | 72 | |
AK057039.1 | - | 22 - 23 | 2707 | 72 | ||
AK127816.1 | - | 10 - 11 | 1399 | 72 | ||
BC021905.1 | - | 22 - 23 | 2654 | 72 | ||
BC137488.1 | - | 22 - 23 | 2645 | 9021488 | 72 | |
DQ924532.1 | - | 22 - 23 | 2539 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |