Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980397 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400692 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400653 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980372 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P17002520 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P17002497 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980379 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400672 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400657 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400688 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980362 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400656 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400707 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400664 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400709 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400714 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400684 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980422 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980406 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400721 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400712 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P17002508 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980361 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400702 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400646 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980415 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P17002498 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980421 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400680 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400676 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980349 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400717 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980384 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980331 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980390 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980348 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P17002506 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400689 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400663 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400678 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400711 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400682 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400720 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400670 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P17002518 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980347 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400703 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400671 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400696 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400686 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400650 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P17002517 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400654 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400719 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400695 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980371 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980334 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980380 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980420 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980342 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980338 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980392 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980343 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400677 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400710 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980346 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10400699 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30980359 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
150557 |
BC034478.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
150557 |
BC093632.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
150557 |
BC110404.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
150557 |
BC111941.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
150557 |
CR611548.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
150557 |
CR612908.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
150557 |
NM_001441.2 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
150557 |
U82535.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
522 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
628445 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
294808 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
178919 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
641334 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P164100 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P164100 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
195 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P128634 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P128634 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
14885 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P34204361 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P34204316 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P03270185 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P20693054 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P20693058 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P164100 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_23_P164100 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400686 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980359 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980380 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980370 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980345 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400671 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P17002508 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980379 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400677 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980349 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400656 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980371 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980343 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P17002506 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400678 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400684 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400676 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980348 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P17002497 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980385 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980347 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980361 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980342 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980362 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400659 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980346 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400682 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400654 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400692 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980384 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400672 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400663 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400689 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400657 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980345 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400677 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P17002497 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P17002506 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400692 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400659 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980379 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980359 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400678 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400657 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980370 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980385 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400676 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400656 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980362 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400663 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980342 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400689 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400682 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400686 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400671 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980384 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400684 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P17002508 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980361 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980371 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400654 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980349 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980346 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980343 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980347 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980348 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30980380 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10400672 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_23_P164100 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P128634 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10400677 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10400676 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10400661 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10400667 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10400666 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10400672 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10400674 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10400665 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10400657 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10400654 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10400663 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10400678 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10400669 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10400671 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10400659 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P17002500 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P17002499 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P17002502 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P10400670 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P17002501 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P17002498 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P17002497 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P10400671 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P20693067 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P128634 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P128634 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P164100 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
133904 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
31870 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P20693067 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P128634 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P20693054 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P20693058 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P03270184 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400646 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400674 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400671 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400653 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400661 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P17002508 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400676 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400711 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P17002501 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P17002497 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400663 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400692 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400695 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400648 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P17002506 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400703 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400665 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400666 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400679 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400694 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400672 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400713 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400686 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P17002529 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400659 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400651 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400707 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400657 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400699 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400706 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400684 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400719 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400709 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400677 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400701 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400690 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400689 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400712 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400715 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400678 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10400721 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P20693067 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P128634 |
- |
- |