Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000447046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000447046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000447046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000469114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000469114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000469114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000469114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000490360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000490360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000490360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000447046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000447046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000447046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000469114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000469114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000469114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000469114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000490360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000490360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000490360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156207.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156207.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156207.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156207.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156207.hg.1 |
ENST00000257632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156207.hg.1 |
ENST00000334348.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156207.hg.1 |
ENST00000394849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156207.hg.1 |
NM_182684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156207.hg.1 |
TC0700008096.hg.1 |
Clariom D TC |