Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003044.hg.1 |
ENST00000406438.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003044.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003044.hg.1 |
TC17000210.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007116.hg.1 |
ENST00000406438.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007116.hg.1 |
ENST00000406438.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007116.hg.1 |
ENST00000406438.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007116.hg.1 |
ENST00000406438.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007116.hg.1 |
ENST00000406438.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007116.hg.1 |
ENST00000406438.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007116.hg.1 |
ENST00000406438.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007116.hg.1 |
ENST00000577764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007116.hg.1 |
ENST00000577764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007116.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007116.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007116.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007116.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007116.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007116.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007116.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007116.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003043.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003043.hg.1 |
TC17000210.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003042.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003042.hg.1 |
TC17000210.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700150283.hg.1 |
ENST00000406438.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700150283.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700150283.hg.1 |
ENST00000406438.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700150283.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700150283.hg.1 |
TC1700007116.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700007116.hg.1 |
ENST00000406438.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007116.hg.1 |
ENST00000406438.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007116.hg.1 |
ENST00000406438.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007116.hg.1 |
ENST00000406438.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007116.hg.1 |
ENST00000406438.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007116.hg.1 |
ENST00000406438.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007116.hg.1 |
ENST00000406438.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007116.hg.1 |
ENST00000577764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007116.hg.1 |
ENST00000577764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007116.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007116.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007116.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007116.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007116.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007116.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007116.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007116.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700150281.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700150281.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700150281.hg.1 |
TC1700007116.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700150282.hg.1 |
ENST00000406438.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700150282.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700150282.hg.1 |
ENST00000406438.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700150282.hg.1 |
NM_144775.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700150282.hg.1 |
TC1700007116.hg.1 |
Clariom D TC |