Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000599871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000599871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000599871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000599871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000599871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000602105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000602105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
ENST00000602105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000332955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000595937.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000597553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000599871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000599871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000599871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000599871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000599871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000602105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000602105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
ENST00000602105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011095.hg.1 |
NM_182575.2 |
Refseq |