Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015725.hg.1 |
TC06001313.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015725.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015725.hg.1 |
ENST00000541730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015725.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174548.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174548.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174548.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174548.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174548.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174548.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174548.hg.1 |
TC0600010991.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
ENST00000324607.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NM_001080480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010991.hg.1 |
NR_073465.1 |
Refseq |