Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000360668.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000360668.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000360668.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000360668.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000360668.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000360668.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000360668.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000360668.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000360668.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000397709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000397709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000397709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000397709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000397709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000397709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000397709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000476428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000476428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000476428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000476428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000476428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
ENST00000476428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
NM_032514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
NM_032514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
NM_032514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
NM_032514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
NM_032514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
NM_032514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
NM_032514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
NM_032514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007193.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003097.hg.1 |
ENST00000374837.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003097.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003097.hg.1 |
TC20000243.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003098.hg.1 |
ENST00000374837.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003098.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003098.hg.1 |
TC20000243.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003104.hg.1 |
ENST00000360668.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003104.hg.1 |
ENST00000374837.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003104.hg.1 |
ENST00000397709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003104.hg.1 |
NM_032514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003104.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003104.hg.1 |
TC20000243.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000149365.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149365.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149365.hg.1 |
TC2000007193.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000149365.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149365.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000360668.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000360668.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000360668.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000360668.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000360668.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000360668.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000360668.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000360668.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000360668.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000397709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000397709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000397709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000397709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000397709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000397709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000397709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000476428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000476428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000476428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000476428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000476428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
ENST00000476428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
NM_032514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
NM_032514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
NM_032514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
NM_032514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
NM_032514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
NM_032514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
NM_032514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
NM_032514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007193.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149367.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149367.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149367.hg.1 |
TC2000007193.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000149367.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149367.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149366.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149366.hg.1 |
NM_181509.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149366.hg.1 |
TC2000007193.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000149366.hg.1 |
ENST00000374837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149366.hg.1 |
NM_181509.2 |
Refseq |