Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006075.hg.1 |
ENST00000291552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006075.hg.1 |
ENST00000398137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006075.hg.1 |
ENST00000459639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006075.hg.1 |
NM_001025204.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006075.hg.1 |
NM_006758.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006075.hg.1 |
TC21000501.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100151044.hg.1 |
ENST00000610664.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100151044.hg.1 |
ENST00000620065.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100151044.hg.1 |
ENST00000623375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100151044.hg.1 |
ENST00000624739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100151044.hg.1 |
NM_001025204.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100151044.hg.1 |
NM_006758.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100151044.hg.1 |
TC2100007492.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100151044.hg.1 |
ENST00000610664.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100151044.hg.1 |
ENST00000620065.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100151044.hg.1 |
ENST00000623375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100151044.hg.1 |
ENST00000624739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100151044.hg.1 |
NM_001025204.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100151044.hg.1 |
NM_006758.2 |
Refseq |