Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005332.2 | NP_005323.1 | 3 | 478 | 148 | |
XM_005255287.3 | XP_005255344.1 | 4 | 1108 | 148 | ||
XM_005255288.3 | XP_005255345.1 | 4 | 1225 | 148 | ||
GenBank mRNA | BC027892.1 | - | 3 | 458 | 5224569 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Human Stem Cell Pluripotency 1700 Inventoried Human_Stem_Cell Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30386617 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16835964 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30386621 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09880741 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30386598 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30386593 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09880736 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30386597 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09880738 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09880732 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16835987 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09880729 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16835966 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30386595 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09880731 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09880739 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09880740 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3252534 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3421827 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P3651 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 10200 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 14056 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 53573 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 12849 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P03100112 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3252534 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3421827 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3252534 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3421827 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30386595 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09880731 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09880729 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30386598 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09880736 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16835964 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09880740 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30386593 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09880741 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30386597 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09880738 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16835966 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09880739 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09880739 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09880741 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30386598 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09880738 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16835966 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16835964 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30386597 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09880731 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30386595 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09880729 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09880736 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09880740 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30386593 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P3651 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09880739 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09880734 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09880736 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09880738 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09880741 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09880740 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P09880736 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P09880739 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P09880742 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P09880740 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P09880741 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P09880734 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P09880738 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P09880739 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P09880740 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16835984 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P09880741 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16835985 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P09880735 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P16835962 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P16835964 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P09880741 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P09880734 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P16835986 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P09880742 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P16835965 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P09880737 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P09880736 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P09880740 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P09880739 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P16835987 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P09880738 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P09880733 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P16835984 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P16835966 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P16835985 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P16835961 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P16835963 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P3651 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03100112 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09880755 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09880728 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09880740 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09880752 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09880741 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09880749 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09880758 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16835959 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16835964 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09880756 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09880739 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09880748 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09880754 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09880738 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09880724 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09880729 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09880731 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16835957 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09880734 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09880736 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16835987 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16835962 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09880726 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_005332_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7991755 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7991756 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7991752 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7991751 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7991753 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7991754 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7991750 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 206647_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11726129_at | - | - |
Affymetrix Human Genome U219 Array | 11726129_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1600011312.hg.1 | ENST00000252951.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011312.hg.1 | ENST00000252951.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011312.hg.1 | ENST00000252951.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011312.hg.1 | ENST00000252951.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011312.hg.1 | ENST00000252951.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011312.hg.1 | ENST00000252951.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011312.hg.1 | NM_005332.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011312.hg.1 | NM_005332.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011312.hg.1 | NM_005332.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011312.hg.1 | NM_005332.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011312.hg.1 | NM_005332.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600188407.hg.1 | ENST00000252951.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188407.hg.1 | NM_005332.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600188407.hg.1 | ENST00000252951.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188407.hg.1 | NM_005332.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600188407.hg.1 | TC1600011312.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600011312.hg.1 | ENST00000252951.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011312.hg.1 | ENST00000252951.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011312.hg.1 | ENST00000252951.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011312.hg.1 | ENST00000252951.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011312.hg.1 | ENST00000252951.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011312.hg.1 | ENST00000252951.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011312.hg.1 | NM_005332.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011312.hg.1 | NM_005332.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011312.hg.1 | NM_005332.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011312.hg.1 | NM_005332.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011312.hg.1 | NM_005332.2 | Refseq |
Human Genome U133 Plus 2.0 Array | 206647_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 206647_at | - | - |
Human Genome U133A 2.0 Array | 206647_PM_at | - | - |
Human Genome U133A 2.0 Array | 206647_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_005332_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw525350 | - | - |
Human Genome U95 Set | 37217_PM_at | - | - |
Human Genome U95 Set | 37217_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 206647_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 206647_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 206647_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 206647_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 206647_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 206647_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 206647_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 206647_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 206647_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 206647_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 206647_at2 | - | - |
Human Genome U133 Set | 206647_at | - | - |
Human Genome U133 Set | 206647_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054098 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054098 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 144454 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1713458 | NM_005332.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1713458 | BC027892.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1713458 | NM_005332.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1713458 | BC027892.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1713458 | NM_005332.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1713458 | BC027892.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1713458 | NM_005332.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1713458 | BC027892.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1713458 | NM_005332.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1713458 | BC027892.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_6633805-S | NM_005332.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_6633805-S | BC027892.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_19775-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0005270021 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0005270021 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1713458 | NM_005332.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1713458 | BC027892.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1713458 | NM_005332.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1713458 | BC027892.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 5720372 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_19775 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1713458 | NM_005332.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1713458 | BC027892.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1713458 | NM_005332.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1713458 | BC027892.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1713458 | NM_005332.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1713458 | BC027892.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1713458 | NM_005332.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1713458 | BC027892.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1713458 | NM_005332.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1713458 | BC027892.1 | human_transcriptome |