Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000492378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000492378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000492378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000492378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000492378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000492378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000494627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000494627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000494627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000494627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000496461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000496461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000496461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000496461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000496461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000496461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000496461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000496461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000496461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000564400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000564400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000564400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000564400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000564400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000564400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000564400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000564400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000564400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000568368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000568368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000568368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000568368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000568368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
ENST00000568368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000245206.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000434819.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000492378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000492378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000492378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000492378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000492378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000492378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000494627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000494627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000494627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000494627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000496461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000496461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000496461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000496461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000496461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000496461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000496461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000496461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000496461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000564400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000564400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000564400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000564400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000564400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000564400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000564400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000564400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000564400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000568368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000568368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000568368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000568368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000568368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
ENST00000568368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_001286220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010500.hg.1 |
NM_002080.3 |
Refseq |