Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000609375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000609375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000609375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000609375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
ENST00000609375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NR_072977.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NR_072977.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NR_072977.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NR_072977.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NR_072977.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NR_072977.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NR_072977.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008379.hg.1 |
NR_072977.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008892.hg.1 |
ENST00000347364.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008892.hg.1 |
ENST00000399054.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008892.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008892.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008892.hg.1 |
TC20000619.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000158981.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158981.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158981.hg.1 |
ENST00000609375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158981.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158981.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158981.hg.1 |
TC2000008379.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000158981.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158981.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158981.hg.1 |
ENST00000609375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158981.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158981.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000609375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000609375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000609375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000609375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
ENST00000609375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NR_072977.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NR_072977.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NR_072977.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NR_072977.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NR_072977.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NR_072977.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NR_072977.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008379.hg.1 |
NR_072977.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158983.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158983.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158983.hg.1 |
ENST00000609375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158983.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158983.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158983.hg.1 |
TC2000008379.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000158983.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158983.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158983.hg.1 |
ENST00000609375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158983.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158983.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158979.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158979.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158979.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158979.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158979.hg.1 |
TC2000008379.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000158979.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158979.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158979.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158979.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158982.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158982.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158982.hg.1 |
ENST00000609375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158982.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158982.hg.1 |
NM_170784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158982.hg.1 |
TC2000008379.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000158982.hg.1 |
ENST00000347364.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158982.hg.1 |
ENST00000399054.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158982.hg.1 |
ENST00000609375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158982.hg.1 |
NM_018848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158982.hg.1 |
NM_170784.2 |
Refseq |