Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021087.hg.1 |
TC02001351.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021087.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021087.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000488477.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000488477.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000488477.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000488477.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021085.hg.1 |
TC02001351.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021085.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021085.hg.1 |
ENST00000488477.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021085.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021083.hg.1 |
TC02001351.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021083.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021083.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200178250.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200178250.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200178250.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200178250.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200178250.hg.1 |
TC0200010911.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200178249.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200178249.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200178249.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200178249.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200178249.hg.1 |
TC0200010911.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000488477.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000488477.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000488477.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000488477.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200178252.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200178252.hg.1 |
ENST00000488477.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200178252.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200178252.hg.1 |
ENST00000281830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200178252.hg.1 |
ENST00000488477.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200178252.hg.1 |
NM_080671.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200178252.hg.1 |
TC0200010911.hg.1 |
Clariom D TC |