Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000498093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000498093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000498093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
ENST00000498093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02012944.hg.1 |
TC02000877.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02012944.hg.1 |
ENST00000283054.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02012944.hg.1 |
ENST00000356140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02012944.hg.1 |
ENST00000392928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02012944.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200165890.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200165890.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200165890.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200165890.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200165890.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200165890.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200165890.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200165890.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200165890.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200165890.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200165890.hg.1 |
TC0200009424.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000356140.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000392928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000485893.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000498093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000498093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000498093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
ENST00000498093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_001307983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009424.hg.1 |
NM_138326.2 |
Refseq |