Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152765.3 | NP_689978.2 | 4 - 5 | 740 | 79 | |
GenBank mRNA | AK055047.1 | - | 1 - 2 | 74 | 79 | |
AK096336.1 | - | 5 - 6 | 528 | 79 | ||
AK308800.1 | - | 5 - 6 | 567 | 79 | ||
AK315486.1 | - | 4 - 5 | 483 | 79 | ||
BC028400.2 | - | 4 - 5 | 464 | 4816822 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |