Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
ENST00000504618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
ENST00000504618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
ENST00000504618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
ENST00000504618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009330.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011747.hg.1 |
TC04000856.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011747.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011747.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163993.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163993.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163993.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163993.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163993.hg.1 |
TC0400009330.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400163994.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163994.hg.1 |
ENST00000504618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163994.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163994.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163994.hg.1 |
ENST00000504618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163994.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163994.hg.1 |
TC0400009330.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400163996.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163996.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163996.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163996.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163996.hg.1 |
TC0400009330.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400163995.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163995.hg.1 |
ENST00000504618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163995.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163995.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163995.hg.1 |
ENST00000504618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163995.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163995.hg.1 |
TC0400009330.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
ENST00000296504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
ENST00000504618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
ENST00000504618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
ENST00000504618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
ENST00000504618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
NM_003864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009330.hg.1 |
NM_003864.3 |
Refseq |