Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007884.hg.1 |
TC04000542.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007884.hg.1 |
ENST00000305737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007884.hg.1 |
ENST00000394764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007884.hg.1 |
NM_017628.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007883.hg.1 |
TC04000542.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007883.hg.1 |
ENST00000305737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007883.hg.1 |
ENST00000394764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007883.hg.1 |
NM_017628.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400157110.hg.1 |
ENST00000305737.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400157110.hg.1 |
NM_017628.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400157110.hg.1 |
ENST00000305737.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400157110.hg.1 |
NM_017628.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400157110.hg.1 |
TC0400008318.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400157111.hg.1 |
ENST00000305737.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400157111.hg.1 |
NM_017628.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400157111.hg.1 |
ENST00000305737.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400157111.hg.1 |
NM_017628.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400157111.hg.1 |
TC0400008318.hg.1 |
Clariom D TC |