Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014427.hg.1 |
ENST00000322357.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014427.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014427.hg.1 |
TC19001057.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014426.hg.1 |
ENST00000322357.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014426.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014426.hg.1 |
TC19001057.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014425.hg.1 |
ENST00000322357.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014425.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014425.hg.1 |
TC19001057.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014430.hg.1 |
ENST00000322357.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014430.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014430.hg.1 |
TC19001057.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014431.hg.1 |
ENST00000322357.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014431.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014431.hg.1 |
TC19001057.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014428.hg.1 |
ENST00000322357.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014428.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014428.hg.1 |
TC19001057.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
ENST00000600056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
ENST00000600056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
ENST00000601588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
ENST00000601588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
ENST00000601588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
ENST00000601588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009320.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900174441.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174441.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900174441.hg.1 |
TC1900009320.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900174441.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174441.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900174439.hg.1 |
ENST00000601588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174439.hg.1 |
TC1900009320.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900174439.hg.1 |
ENST00000601588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
ENST00000600056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
ENST00000600056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
ENST00000601588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
ENST00000601588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
ENST00000601588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
ENST00000601588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009320.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900174430.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174430.hg.1 |
ENST00000601588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174430.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900174430.hg.1 |
TC1900009320.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900174430.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174430.hg.1 |
ENST00000601588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174430.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900174432.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174432.hg.1 |
ENST00000601588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174432.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900174432.hg.1 |
TC1900009320.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900174432.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174432.hg.1 |
ENST00000601588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174432.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900174442.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174442.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900174442.hg.1 |
TC1900009320.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900174442.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174442.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900174426.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174426.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900174426.hg.1 |
TC1900009320.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900174426.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174426.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900174427.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174427.hg.1 |
NM_015898.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900174427.hg.1 |
TC1900009320.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900174427.hg.1 |
ENST00000322357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174427.hg.1 |
NM_015898.3 |
Refseq |