Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
ENST00000444131.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
ENST00000457408.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
ENST00000477639.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
ENST00000481238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
NM_001303058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
NM_001303060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
NM_001303061.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
NM_001303062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
NM_182547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
ENST00000444131.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
ENST00000457408.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
ENST00000477639.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
ENST00000481238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
NM_001303058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
NM_001303060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
NM_001303061.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
NM_001303062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
NM_182547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178039.hg.1 |
TC0700010926.hg.1 |
Clariom D TC |