Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056199.hg.1 |
TC01003663.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056199.hg.1 |
ENST00000367405.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056199.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056199.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016833.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227403.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227403.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227403.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227403.hg.1 |
ENST00000367405.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227403.hg.1 |
ENST00000467322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227403.hg.1 |
NM_194314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227403.hg.1 |
TC0100016833.hg.1 |
Clariom D TC |