Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC007888.2 | - | 1 | 41 | 4120898 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens 1700 Inventoried Amplicon spans exons and probe does not span exons |