Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK127052.1 | - | 1 | 3051 | 109 |
Gene Symbol: LOC105370939 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK093758.1 | - | 4 | 1266 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564974 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564941 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565014 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30051696 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30051767 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30051690 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564948 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564940 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16772018 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564960 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564933 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564943 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564977 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30051772 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16772043 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565007 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564985 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16772036 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30051761 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16772042 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30051773 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564993 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565004 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565012 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564950 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16772037 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565003 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564952 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564969 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565000 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565013 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565001 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564954 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564992 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565010 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564939 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564982 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16772041 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30051759 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565008 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564961 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30051752 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30051714 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564962 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564991 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564986 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564997 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564979 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30051758 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30051724 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565006 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564955 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30051725 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564988 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564971 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564968 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564951 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564989 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30051751 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30051775 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564967 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564953 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564945 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564966 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564932 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564957 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564959 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564947 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564973 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564935 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30051749 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09564946 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30051754 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 56930 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 2736 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 334096 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 517771 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 809789 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 766384 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 278081 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 413372 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3336592 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P118326 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P118326 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 29714 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_14_P118326 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3336592 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P118326 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P20200306 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P118326 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P118326 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 89927 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 9192 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03001086 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P118326 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03001079 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20200306 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P845595 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P118326 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P20200306 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7982334 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7982333 | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000500533.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000500533.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000500533.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000500533.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000500533.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000500533.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000558225.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000558225.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000558225.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000558225.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000558673.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000558673.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000558844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000558844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000558921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000558921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000560473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000560473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000560473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000560473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000618680.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000618680.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000618680.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000618680.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000618680.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006756.hg.1 | ENST00000618680.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000307145.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000500533.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000500533.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000500533.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000500533.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000500533.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000500533.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000558225.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000558225.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000558225.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000558225.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000558673.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000558673.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000558844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000558844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000558921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000558921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000560473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000560473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000560473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000560473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000618680.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000618680.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000618680.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000618680.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000618680.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006756.hg.1 | ENST00000618680.4 | Ensembl |
Human Genome U133 Plus 2.0 Array | 1564463_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1564463_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564463_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564463_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564463_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564463_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564463_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564463_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564463_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564463_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564463_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564463_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564463_at8 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 541734 | - | - |