Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013897.hg.1 |
ENST00000216241.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013897.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013897.hg.1 |
TC22000822.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013899.hg.1 |
ENST00000216241.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013899.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013899.hg.1 |
TC22000822.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000455425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000455425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000455425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000455425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000455425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
ENST00000455425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200163840.hg.1 |
TC2200008818.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200163840.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163840.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163840.hg.1 |
ENST00000455425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163840.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200163840.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163840.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163840.hg.1 |
ENST00000455425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163840.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200163837.hg.1 |
TC2200008818.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200163837.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163837.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163837.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200163837.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163837.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163837.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200163838.hg.1 |
TC2200008818.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200163838.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163838.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163838.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200163838.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163838.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163838.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000216241.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000417999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000455425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000455425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000455425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000455425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000455425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
ENST00000455425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008818.hg.1 |
NM_138481.1 |
Refseq |