Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008840.hg.1 |
TC02000578.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008840.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000272610.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000272610.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000272610.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000272610.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000272610.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000272610.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000272610.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000272610.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000272610.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000272610.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000414820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000414820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000414820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000414820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000414820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000414820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000414820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000414820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000414820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000414820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000463096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000463096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000463096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000463096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000463096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000463096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000463096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000463096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000468548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000468548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000468548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000474849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000474849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000474849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000474849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000474849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000483657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000483657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000483657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
ENST00000483657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
NM_199336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
NM_199336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
NM_199336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
NM_199336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
NM_199336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
NM_199336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
NM_199336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
NM_199336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013552.hg.1 |
NM_199336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158877.hg.1 |
ENST00000443258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158877.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158877.hg.1 |
ENST00000483000.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158877.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158877.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158877.hg.1 |
ENST00000443258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158877.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158877.hg.1 |
ENST00000483000.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158877.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158877.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158877.hg.1 |
TC0200008493.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158875.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158875.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158875.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158875.hg.1 |
ENST00000467292.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158875.hg.1 |
ENST00000607780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158875.hg.1 |
NR_003698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158875.hg.1 |
TC0200008493.hg.1 |
Clariom D TC |