Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000397428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000397428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000397428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000397428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000503121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000503121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000503121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000503121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000503121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000505054.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000505054.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000505054.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000505054.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000505054.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
ENST00000505054.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019733.hg.1 |
TC19002696.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019733.hg.1 |
ENST00000324444.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019733.hg.1 |
ENST00000340477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019733.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000397428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000397428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000397428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000397428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000503121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000503121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000503121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000503121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000503121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000505054.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000505054.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000505054.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000505054.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000505054.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
ENST00000505054.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011931.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900205706.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205706.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205706.hg.1 |
ENST00000397428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205706.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205706.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205706.hg.1 |
ENST00000505054.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205706.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900205706.hg.1 |
NM_001297735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900205706.hg.1 |
TC1900011931.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900205706.hg.1 |
ENST00000324444.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205706.hg.1 |
ENST00000340477.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205706.hg.1 |
ENST00000397428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205706.hg.1 |
ENST00000465425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205706.hg.1 |
ENST00000490730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205706.hg.1 |
ENST00000505054.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205706.hg.1 |
NM_001039876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900205706.hg.1 |
NM_001297735.2 |
Refseq |