Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000444116.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000444116.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000444116.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000444116.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000444116.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000456164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000456164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000456164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000456164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000456164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000478089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000478089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000478089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
ENST00000478089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000444116.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000444116.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000444116.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000444116.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000444116.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000456164.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000456164.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000456164.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000456164.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000456164.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000478089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000478089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000478089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
ENST00000478089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013038.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196113.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196113.hg.1 |
ENST00000444116.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196113.hg.1 |
ENST00000456164.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196113.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196113.hg.1 |
ENST00000375592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196113.hg.1 |
ENST00000444116.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196113.hg.1 |
ENST00000456164.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196113.hg.1 |
NM_018994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196113.hg.1 |
TC0100013038.hg.1 |
Clariom D TC |