Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
ENST00000409911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001282928.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001282928.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001282928.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001282928.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001282928.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001282928.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_001282928.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016693.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032325.hg.1 |
TC02002011.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032325.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032325.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032325.hg.1 |
ENST00000409282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032325.hg.1 |
ENST00000409884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032325.hg.1 |
ENST00000409911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032325.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032325.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032326.hg.1 |
TC02002011.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032326.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032326.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032326.hg.1 |
ENST00000409282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032326.hg.1 |
ENST00000409884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032326.hg.1 |
ENST00000409911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032326.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032326.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
ENST00000409282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
ENST00000409884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
ENST00000409911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
NM_001282928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
ENST00000409282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
ENST00000409884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
ENST00000409911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
NM_001282928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226988.hg.1 |
TC0200016693.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409088.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
ENST00000409911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001134745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001282924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001282928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001282928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001282928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001282928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001282928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001282928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_001282928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_024993.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016693.hg.1 |
NM_024993.5 |
Refseq |