Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012065.hg.1 |
ENST00000300399.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012065.hg.1 |
ENST00000397452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012065.hg.1 |
ENST00000432451.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012065.hg.1 |
ENST00000534972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012065.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012065.hg.1 |
TC22000683.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200161331.hg.1 |
TC2200008515.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200161331.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161331.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161331.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161331.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161331.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161331.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161331.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161331.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000300399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000397452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
ENST00000534972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008515.hg.1 |
NM_174932.2 |
Refseq |