Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL5A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000393.3 | NP_000384.2 | 18 - 19 | 1436 | 75 | |
XM_011510573.2 | XP_011508875.1 | 18 - 19 | 1218 | 75 | ||
GenBank mRNA | AK297936.1 | - | 18 - 19 | 1318 | 75 | |
J04478.1 | - | 18 - 19 | 1299 | 75 | ||
Y14690.1 | - | 18 - 19 | 1299 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |