Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035885.hg.1 |
TC03002011.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035885.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035885.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010507.hg.1 |
ENST00000556538.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010507.hg.1 |
ENST00000556538.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198296.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198296.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198296.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198296.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198296.hg.1 |
TC0300013162.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
ENST00000351008.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013162.hg.1 |
NM_031955.5 |
Refseq |