Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004057.hg.1 |
TC02000272.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004057.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004057.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007402.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200151274.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151274.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200151274.hg.1 |
ENST00000272286.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151274.hg.1 |
NM_022437.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200151274.hg.1 |
TC0200007402.hg.1 |
Clariom D TC |