Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEDD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142393.1 | NP_001135865.1 | 3 - 4 | 815 | 147 | |
NM_006403.3 | NP_006394.1 | 2 - 3 | 620 | 147 | ||
GenBank mRNA | AK292682.1 | - | 3 - 4 | 815 | 147 | |
BC040207.1 | - | 2 - 3 | 606 | 4820745 | 147 | |
L43821.1 | - | 2 - 3 | 616 | 147 | ||
U64317.1 | - | 2 - 3 | 591 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |