Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083588.1 | NP_001077057.1 | 6 - 7 | 921 | 88 | |
GenBank mRNA | CN312917.1 | - | 3 - 4 | 361 | 88 | |
U15642.1 | - | 6 - 7 | 1087 | 88 | ||
U31556.1 | - | 6 - 7 | 922 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |