Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109763.1 | NP_001103233.1 | 2 - 3 | 480 | 91 | |
NM_001323900.1 | NP_001310829.1 | 2 - 3 | 480 | 91 | ||
XM_011545750.2 | XP_011544052.1 | 2 - 3 | 474 | 91 | ||
XM_011545751.2 | XP_011544053.1 | 2 - 3 | 474 | 91 | ||
GenBank mRNA | AK128775.1 | - | 2 - 3 | 101 | 91 | |
BI752847.1 | - | 2 - 3 | 158 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |