Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027961.hg.1 |
TC01001788.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027961.hg.1 |
ENST00000261454.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027961.hg.1 |
ENST00000366958.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027961.hg.1 |
ENST00000435016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027961.hg.1 |
ENST00000498508.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027961.hg.1 |
ENST00000541470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027961.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000471129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000471129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000471129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000471129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000607425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000607425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000607425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000607726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
ENST00000607726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027960.hg.1 |
TC01001788.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027960.hg.1 |
ENST00000261454.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027960.hg.1 |
ENST00000366958.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027960.hg.1 |
ENST00000435016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027960.hg.1 |
ENST00000498508.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027960.hg.1 |
ENST00000541470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027960.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185195.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185195.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185195.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185195.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185195.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185195.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185195.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185195.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185195.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185195.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185195.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185195.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185195.hg.1 |
TC0100011566.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100185196.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185196.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185196.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185196.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185196.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185196.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185196.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185196.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185196.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185196.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185196.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185196.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185196.hg.1 |
TC0100011566.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100185198.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185198.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185198.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185198.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185198.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185198.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185198.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185198.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185198.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185198.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185198.hg.1 |
TC0100011566.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000261454.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000366958.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000435016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000471129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000471129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000471129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000471129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000498508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000607425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000607425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000607425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000607726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
ENST00000607726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_001270616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011566.hg.1 |
NM_002763.4 |
Refseq |