Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000326524.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000326524.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000326524.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000326524.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000326524.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000326524.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000326524.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000326524.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000344622.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000344622.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000344622.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000344622.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000344622.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000344622.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000344622.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000381826.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000381826.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000381826.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000381826.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000381826.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000381826.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000427982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000427982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000427982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000427982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000427982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000427982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000427982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000502572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000502572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000502572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000502572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000510177.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000510177.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000510177.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000510177.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000515058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000515058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000515058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000515058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000515058.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000620847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000620847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000620847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000620847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000620847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000620847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
ENST00000620847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_000514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_000514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_000514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_000514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_000514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_000514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_000514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_000514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001190469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001190469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001190469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001190469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001190469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001190469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001190469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001190469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001278098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001278098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001278098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001278098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001278098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001278098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_001278098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_199231.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_199231.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_199231.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_199231.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_199231.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_199231.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010523.hg.1 |
NM_199231.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000326524.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000326524.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000326524.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000326524.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000326524.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000326524.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000326524.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000326524.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000344622.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000344622.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000344622.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000344622.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000344622.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000344622.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000344622.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000381826.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000381826.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000381826.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000381826.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000381826.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000381826.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000427982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000427982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000427982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000427982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000427982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000427982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000427982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000502572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000502572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000502572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000502572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000510177.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000510177.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000510177.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000510177.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000515058.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000515058.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000515058.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000515058.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000515058.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000620847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000620847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000620847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000620847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000620847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000620847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
ENST00000620847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_000514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_000514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_000514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_000514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_000514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_000514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_000514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_000514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001190469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001190469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001190469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001190469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001190469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001190469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001190469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001190469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001278098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001278098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001278098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001278098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001278098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001278098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_001278098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_199231.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_199231.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_199231.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_199231.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_199231.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_199231.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010523.hg.1 |
NM_199231.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173354.hg.1 |
ENST00000326524.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173354.hg.1 |
ENST00000344622.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173354.hg.1 |
ENST00000620847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173354.hg.1 |
NM_000514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173354.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173354.hg.1 |
NM_001190469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173354.hg.1 |
NM_001278098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173354.hg.1 |
NM_199231.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173354.hg.1 |
ENST00000326524.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173354.hg.1 |
ENST00000344622.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173354.hg.1 |
ENST00000620847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173354.hg.1 |
NM_000514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173354.hg.1 |
NM_001190468.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173354.hg.1 |
NM_001190469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173354.hg.1 |
NM_001278098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173354.hg.1 |
NM_199231.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173354.hg.1 |
TC0500010523.hg.1 |
Clariom D TC |