Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XRCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021141.3 | NP_066964.1 | 19 - 20 | 2246 | 72 | |
GenBank mRNA | AK026166.1 | - | 14 - 15 | 1482 | 72 | |
AK096408.1 | - | 16 - 17 | 2648 | 72 | ||
AK222603.1 | - | 19 - 20 | 2163 | 72 | ||
AK290740.1 | - | 19 - 20 | 2132 | 72 | ||
BC019027.2 | - | 19 - 20 | 2114 | 4555162 | 72 | |
BC095442.1 | - | 19 - 20 | 2132 | 30329657 | 72 | |
J04977.1 | - | 19 - 20 | 2139 | 72 | ||
M30938.1 | - | 19 - 20 | 2133 | 72 | ||
X57500.1 | - | 16 - 17 | 1795 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |