Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029575.hg.1 |
ENST00000299140.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029575.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029575.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029575.hg.1 |
TC11002468.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012902.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100198843.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100198843.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100198843.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100198843.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100198843.hg.1 |
ENST00000299140.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100198843.hg.1 |
ENST00000532889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100198843.hg.1 |
NM_001291992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100198843.hg.1 |
NM_174927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100198843.hg.1 |
TC1100012902.hg.1 |
Clariom D TC |