Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010799.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015207.hg.1 |
TC06001270.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015207.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015207.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173465.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173465.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173465.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173465.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173465.hg.1 |
TC0600010799.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
ENST00000379491.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
NM_004752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010799.hg.1 |
NM_004752.3 |
Refseq |