Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004187.hg.1 |
ENST00000299824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004187.hg.1 |
ENST00000373331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004187.hg.1 |
ENST00000468265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004187.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004187.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004187.hg.1 |
TC20000291.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000463749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000463749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000463749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000463749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000468265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000468265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000468265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
ENST00000468265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004188.hg.1 |
ENST00000299824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004188.hg.1 |
ENST00000373331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004188.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004188.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004188.hg.1 |
TC20000291.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000150799.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150799.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150799.hg.1 |
ENST00000463749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150799.hg.1 |
ENST00000468265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150799.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150799.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150799.hg.1 |
TC2000007336.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000150799.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150799.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150799.hg.1 |
ENST00000463749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150799.hg.1 |
ENST00000468265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150799.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150799.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000299824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000373331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000463749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000463749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000463749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000463749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000468265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000468265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000468265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
ENST00000468265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_001172735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007336.hg.1 |
NM_015568.3 |
Refseq |