Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011566.hg.1 |
TC01000727.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011566.hg.1 |
ENST00000294428.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011566.hg.1 |
ENST00000371072.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011566.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008607.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100161094.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161094.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161094.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161094.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100161094.hg.1 |
ENST00000294428.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161094.hg.1 |
ENST00000371072.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161094.hg.1 |
ENST00000418058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161094.hg.1 |
NM_018211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100161094.hg.1 |
TC0100008607.hg.1 |
Clariom D TC |