Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170744.4 | NP_734465.2 | 8 - 9 | 1550 | 91 | |
XM_011539453.1 | XP_011537755.1 | 9 - 10 | 1099 | 91 | ||
XM_017015834.1 | XP_016871323.1 | 9 - 10 | 1109 | 91 | ||
XM_017015835.1 | XP_016871324.1 | 8 - 9 | 1902 | 91 | ||
GenBank mRNA | AB096256.1 | - | 8 - 9 | 1505 | 91 | |
AK128132.1 | - | 8 - 9 | 1550 | 91 | ||
AY358351.1 | - | 8 - 9 | 1496 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons |