Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007038.hg.1 |
ENST00000579580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007038.hg.1 |
ENST00000579580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004556.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004556.hg.1 |
ENST00000399218.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004556.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004556.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004556.hg.1 |
TC18000446.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000578619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000578619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000578619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000583696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000583696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
ENST00000583696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157363.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157363.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157363.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157363.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157363.hg.1 |
TC1800008418.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800157363.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157363.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157363.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157363.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007038.hg.1 |
ENST00000579580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007038.hg.1 |
ENST00000579580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000578619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000578619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000578619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000583696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000583696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
ENST00000583696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008418.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157364.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157364.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157364.hg.1 |
ENST00000578619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157364.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157364.hg.1 |
NM_022751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157364.hg.1 |
TC1800008418.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800157364.hg.1 |
ENST00000269209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157364.hg.1 |
ENST00000399218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157364.hg.1 |
ENST00000578619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157364.hg.1 |
NM_001242409.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157364.hg.1 |
NM_022751.2 |
Refseq |