Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14016536.hg.1 |
ENST00000334708.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14016536.hg.1 |
NM_001080451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14016536.hg.1 |
TC14001467.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010047.hg.1 |
ENST00000334708.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010047.hg.1 |
ENST00000334708.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010047.hg.1 |
ENST00000334708.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010047.hg.1 |
ENST00000334708.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010047.hg.1 |
ENST00000334708.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010047.hg.1 |
ENST00000334708.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010047.hg.1 |
ENST00000334708.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010047.hg.1 |
NM_001080451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010047.hg.1 |
NM_001080451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010047.hg.1 |
NM_001080451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010047.hg.1 |
NM_001080451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010047.hg.1 |
NM_001080451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010047.hg.1 |
NM_001080451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010047.hg.1 |
NM_001080451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400174431.hg.1 |
ENST00000334708.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400174431.hg.1 |
NM_001080451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400174431.hg.1 |
ENST00000334708.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400174431.hg.1 |
NM_001080451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400174431.hg.1 |
TC1400010047.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400010047.hg.1 |
ENST00000334708.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010047.hg.1 |
ENST00000334708.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010047.hg.1 |
ENST00000334708.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010047.hg.1 |
ENST00000334708.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010047.hg.1 |
ENST00000334708.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010047.hg.1 |
ENST00000334708.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010047.hg.1 |
ENST00000334708.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010047.hg.1 |
NM_001080451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010047.hg.1 |
NM_001080451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010047.hg.1 |
NM_001080451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010047.hg.1 |
NM_001080451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010047.hg.1 |
NM_001080451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010047.hg.1 |
NM_001080451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010047.hg.1 |
NM_001080451.1 |
Refseq |