Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286775.1 | NP_001273704.1 | 2 - 3 | 210 | 159 | |
NM_138779.4 | NP_620134.3 | 2 - 3 | 210 | 159 | ||
GenBank mRNA | AK314995.1 | - | 2 - 3 | 139 | 159 | |
BC015148.2 | - | 2 - 3 | 181 | 4825712 | 159 | |
BG527702.1 | - | 2 - 3 | 159 | 159 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |