Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000434982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000434982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000434982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000434982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000434982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000434982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000434982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000505970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000505970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000505970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000505970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000505970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000505970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000505970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000513241.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000513241.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000513241.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000513241.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000513241.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000513241.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000513241.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000519114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000519114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000519114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000519114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
ENST00000519114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019161.hg.1 |
TC05001369.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019161.hg.1 |
ENST00000341048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019161.hg.1 |
ENST00000504958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019161.hg.1 |
ENST00000507283.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019161.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175335.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175335.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175335.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175335.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175335.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175335.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175335.hg.1 |
TC0500010795.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000341048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000434982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000434982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000434982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000434982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000434982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000434982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000434982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000504958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000505970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000505970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000505970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000505970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000505970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000505970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000505970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000513241.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000513241.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000513241.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000513241.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000513241.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000513241.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000513241.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000519114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000519114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000519114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000519114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
ENST00000519114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010795.hg.1 |
NM_024669.2 |
Refseq |