Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFSWAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261411.1 | NP_001248340.1 | 7 - 8 | 1223 | 110 | |
NM_004592.3 | NP_004583.2 | 7 - 8 | 1223 | 110 | ||
XM_011538653.1 | XP_011536955.1 | 7 - 8 | 1220 | 110 | ||
XM_011538654.2 | XP_011536956.1 | 7 - 8 | 1175 | 110 | ||
XM_011538655.2 | XP_011536957.1 | 7 - 8 | 1220 | 110 | ||
XM_017019798.1 | XP_016875287.1 | 7 - 8 | 1220 | 110 | ||
XM_017019799.1 | XP_016875288.1 | 7 - 8 | 1662 | 110 | ||
GenBank mRNA | AB208921.1 | - | 5 - 6 | 4414 | 110 | |
BC008707.2 | - | 7 - 8 | 1220 | 110 | ||
BC015953.2 | - | 7 - 8 | 1220 | 110 | ||
BC024041.1 | - | 7 - 8 | 1221 | 110 | ||
BC113420.1 | - | 7 - 8 | 1126 | 110 | ||
BC136678.1 | - | 7 - 8 | 1173 | 9020668 | 110 | |
BC144364.1 | - | 7 - 8 | 1173 | 9052885 | 110 | |
U08377.1 | - | 7 - 8 | 1200 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |