Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018051.4 | NP_060521.4 | 16 - 17 | 2251 | 68 | |
XM_005249549.1 | XP_005249606.1 | 13 - 14 | 1843 | 68 | ||
XM_006716041.2 | XP_006716104.1 | 16 - 17 | 2275 | 68 | ||
XM_011516367.1 | XP_011514669.1 | 16 - 17 | 2117 | 68 | ||
XM_011516368.2 | XP_011514670.1 | 16 - 17 | 2275 | 68 | ||
XM_011516369.2 | XP_011514671.1 | 16 - 17 | 2106 | 68 | ||
XM_017012381.1 | XP_016867870.1 | 16 - 17 | 2297 | 68 | ||
XM_017012382.1 | XP_016867871.1 | 16 - 17 | 2266 | 68 | ||
XM_017012383.1 | XP_016867872.1 | 16 - 17 | 2327 | 68 | ||
XM_017012384.1 | XP_016867873.1 | 13 - 14 | 1834 | 68 | ||
XM_017012385.1 | XP_016867874.1 | 15 - 16 | 2008 | 68 | ||
XM_017012386.1 | XP_016867875.1 | 13 - 14 | 1787 | 68 | ||
XM_017012387.1 | XP_016867876.1 | 16 - 17 | 2127 | 68 | ||
XM_017012388.1 | XP_016867877.1 | 6 - 7 | 1011 | 68 | ||
GenBank mRNA | AK027228.1 | - | 5 - 6 | 608 | 68 | |
BC014491.1 | - | 16 - 17 | 2251 | 4906683 | 68 | |
BM718580.1 | - | 1 - 2 | 74 | 68 | ||
CR627036.1 | - | 11 - 12 | 3893 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |