Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018051.4 | NP_060521.4 | 22 - 23 | 2808 | 113 | |
XM_005249549.1 | XP_005249606.1 | 19 - 20 | 2400 | 113 | ||
XM_006716041.2 | XP_006716104.1 | 22 - 23 | 2832 | 113 | ||
XM_011516367.1 | XP_011514669.1 | 22 - 23 | 2674 | 113 | ||
XM_011516369.2 | XP_011514671.1 | 22 - 23 | 2663 | 113 | ||
XM_017012381.1 | XP_016867870.1 | 22 - 23 | 2854 | 113 | ||
XM_017012383.1 | XP_016867872.1 | 22 - 23 | 2884 | 113 | ||
XM_017012384.1 | XP_016867873.1 | 19 - 20 | 2391 | 113 | ||
XM_017012385.1 | XP_016867874.1 | 21 - 22 | 2565 | 113 | ||
XM_017012386.1 | XP_016867875.1 | 19 - 20 | 2344 | 113 | ||
XM_017012387.1 | XP_016867876.1 | 22 - 23 | 2684 | 113 | ||
XM_017012388.1 | XP_016867877.1 | 12 - 13 | 1568 | 113 | ||
GenBank mRNA | AK001162.1 | - | 6 - 7 | 523 | 113 | |
AK027228.1 | - | 11 - 12 | 1164 | 113 | ||
AL137660.1 | - | 17 - 18 | 1808 | 113 | ||
BC014491.1 | - | 22 - 23 | 2808 | 4906683 | 113 | |
CR627036.1 | - | 17 - 18 | 4450 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |